Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020884.4 | NP_065935.3 | 24 - 25 | 2691 | 100 |
XM_006723840.3 | XP_006723903.1 | 22 - 23 | 2332 | 100 | |
XM_011528941.2 | XP_011527243.1 | 22 - 23 | 2356 | 100 | |
XM_011528947.2 | XP_011527249.1 | 16 - 17 | 2729 | 100 | |
XM_011528948.2 | XP_011527250.1 | 16 - 17 | 1888 | 100 | |
XM_017027986.1 | XP_016883475.1 | 23 - 24 | 2392 | 100 | |
XM_017027987.1 | XP_016883476.1 | 18 - 19 | 1907 | 100 | |
GenBank mRNA | AB040945.2 | - | 22 - 23 | 2400 | 100 |
BC151242.1 | - | 22 - 23 | 2412 | 100 |