Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032865.5 | NP_116254.4 | 9 - 10 | 1904 | 68 |
XM_005257744.1 | XP_005257801.1 | 9 - 10 | 1980 | 68 | |
XM_017025236.1 | XP_016880725.1 | 8 - 9 | 1905 | 68 | |
GenBank mRNA | AF370421.1 | - | 9 - 10 | 1901 | 68 |
AF417488.1 | - | 9 - 10 | 1866 | 68 | |
AK027856.1 | - | 7 - 8 | 1254 | 68 | |
BC018706.1 | - | 8 - 9 | 1350 | 68 |