Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010986.2 | NP_001010986.1 | 17 - 18 | 1881 | 124 |
NM_173694.4 | NP_775965.2 | 17 - 18 | 1881 | 124 | |
XM_017029439.1 | XP_016884928.1 | 18 - 19 | 1887 | 124 | |
XM_017029440.1 | XP_016884929.1 | 18 - 19 | 2558 | 124 | |
XM_017029441.1 | XP_016884930.1 | 19 - 20 | 1967 | 124 | |
XM_017029442.1 | XP_016884931.1 | 18 - 19 | 2108 | 124 | |
XM_017029443.1 | XP_016884932.1 | 18 - 19 | 1886 | 124 | |
XM_017029444.1 | XP_016884933.1 | 17 - 18 | 2498 | 124 | |
XM_017029445.1 | XP_016884934.1 | 18 - 19 | 1888 | 124 | |
XM_017029446.1 | XP_016884935.1 | 17 - 18 | 1818 | 124 | |
XM_017029447.1 | XP_016884936.1 | 18 - 19 | 2091 | 124 | |
XM_017029448.1 | XP_016884937.1 | 18 - 19 | 2223 | 124 | |
XM_017029449.1 | XP_016884938.1 | 17 - 18 | 1829 | 124 | |
GenBank mRNA | AJ580093.1 | - | 17 - 18 | 1883 | 124 |
AJ580094.1 | - | 17 - 18 | 1883 | 124 | |
AK125986.1 | - | 7 - 8 | 871 | 124 | |
AK131262.1 | - | 2 - 3 | 157 | 124 | |
BC132728.1 | - | 17 - 18 | 1819 | 124 | |
BC132730.1 | - | 17 - 18 | 1819 | 124 | |
BX648320.1 | - | 4 - 5 | 323 | 124 |