Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031415.2 | NP_113603.1 | 12 - 13 | 2094 | 76 |
XM_011517158.1 | XP_011515460.1 | 11 - 12 | 1807 | 76 | |
XM_011517159.1 | XP_011515461.1 | 10 - 11 | 1777 | 76 | |
XM_011517161.1 | XP_011515463.1 | 12 - 13 | 1834 | 76 | |
XM_011517162.1 | XP_011515464.1 | 12 - 13 | 1834 | 76 | |
XM_011517163.2 | XP_011515465.1 | 11 - 12 | 1061 | 76 | |
XM_011517165.1 | XP_011515467.1 | 7 - 8 | 573 | 76 | |
XM_011517166.2 | XP_011515468.1 | 8 - 9 | 2253 | 76 | |
XM_017013644.1 | XP_016869133.1 | 12 - 13 | 1834 | 76 | |
XM_017013645.1 | XP_016869134.1 | 10 - 11 | 867 | 76 | |
GenBank mRNA | AB042405.1 | - | 12 - 13 | 2103 | 76 |
AJ245876.1 | - | 6 - 7 | 557 | 76 | |
BC035321.1 | - | 12 - 13 | 1639 | 76 | |
BC063595.1 | - | 12 - 13 | 1622 | 76 |