Assay Design



Gene Transcripts

Gene Symbol
GSDMC
Entrez Gene ID
56169
Gene Name
gasdermin C
Gene Aliases
MLZE
Chromosome Location
Chr.8: 129705903 - 129786888 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031415.2 NP_113603.1 13 - 14 2169 98
XM_011517158.1 XP_011515460.1 12 - 13 1882 98
XM_011517159.1 XP_011515461.1 11 - 12 1852 98
XM_011517163.2 XP_011515465.1 12 - 13 1136 98
XM_011517165.1 XP_011515467.1 8 - 9 648 98
XM_011517166.2 XP_011515468.1 9 - 10 2328 98
XM_017013645.1 XP_016869134.1 11 - 12 942 98
GenBank mRNA AB042405.1 - 13 - 14 2178 98
AJ245876.1 - 7 - 8 632 98
BC035321.1 - 13 - 14 1714 98
BC063595.1 - 13 - 14 1697 98

View Full Product Details