Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031415.2 | NP_113603.1 | 4 - 5 | 1459 | 105 |
XM_011517158.1 | XP_011515460.1 | 4 - 5 | 1199 | 105 | |
XM_011517159.1 | XP_011515461.1 | 4 - 5 | 1199 | 105 | |
XM_011517161.1 | XP_011515463.1 | 4 - 5 | 1199 | 105 | |
XM_011517162.1 | XP_011515464.1 | 4 - 5 | 1199 | 105 | |
XM_011517163.2 | XP_011515465.1 | 3 - 4 | 426 | 105 | |
XM_011517167.1 | XP_011515469.1 | 4 - 5 | 1199 | 105 | |
XM_011517168.1 | XP_011515470.1 | 4 - 5 | 1199 | 105 | |
XM_017013644.1 | XP_016869133.1 | 4 - 5 | 1199 | 105 | |
GenBank mRNA | AB042405.1 | - | 4 - 5 | 1468 | 105 |
BC035321.1 | - | 4 - 5 | 1004 | 105 | |
BC063595.1 | - | 4 - 5 | 987 | 105 |