Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003327.3 | NP_003318.1 | 2 - 3 | 308 | 66 |
XM_011542075.2 | XP_011540377.1 | 2 - 3 | 775 | 66 | |
XM_017002231.1 | XP_016857720.1 | 3 - 4 | 1653 | 66 | |
XM_017002232.1 | XP_016857721.1 | 3 - 4 | 1653 | 66 | |
GenBank mRNA | BC105070.1 | - | 2 - 3 | 308 | 66 |
BC105072.1 | - | 2 - 3 | 308 | 66 | |
S76792.1 | - | 2 - 3 | 258 | 66 |