Assay Design



Gene Transcripts

Gene Symbol
SGCZ
Entrez Gene ID
137868
Gene Name
sarcoglycan zeta
Gene Aliases
ZSG1
Chromosome Location
Chr.8: 14084835 - 15238283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322880.1 NP_001309809.1 3 - 4 999 125
NM_001322881.1 NP_001309810.1 3 - 4 905 125
NM_139167.3 NP_631906.2 3 - 4 999 125
GenBank mRNA AY028700.1 - 2 - 3 300 125
AY502063.1 - 3 - 4 339 125
BC069459.1 - 2 - 3 300 125

View Full Product Details