Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322879.1 | NP_001309808.1 | 4 - 5 | 1102 | 84 |
NM_139167.3 | NP_631906.2 | 5 - 6 | 1204 | 84 | |
GenBank mRNA | AY028700.1 | - | 4 - 5 | 505 | 84 |
AY502063.1 | - | 5 - 6 | 544 | 84 | |
BC069459.1 | - | 4 - 5 | 505 | 84 | |
BC125037.1 | - | 3 - 4 | 403 | 84 |