Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007024.1 | NP_001007025.1 | 3 - 4 | 285 | 94 |
NM_001007025.1 | NP_001007026.1 | 3 - 4 | 263 | 94 | |
NM_004871.2 | NP_004862.1 | 3 - 4 | 269 | 94 | |
XM_005258070.4 | XP_005258127.1 | 3 - 4 | 510 | 94 | |
XM_005258071.4 | XP_005258128.1 | 3 - 4 | 503 | 94 | |
XM_005258072.2 | XP_005258129.1 | 3 - 4 | 297 | 94 | |
XM_017025374.1 | XP_016880863.1 | 3 - 4 | 513 | 94 | |
XM_017025375.1 | XP_016880864.1 | 3 - 4 | 272 | 94 | |
XM_017025376.1 | XP_016880865.1 | 3 - 4 | 254 | 94 | |
XM_017025377.1 | XP_016880866.1 | 3 - 4 | 519 | 94 | |
GenBank mRNA | AF047438.1 | - | 3 - 4 | 269 | 94 |
AF073926.1 | - | 3 - 4 | 255 | 94 | |
AK000606.1 | - | 3 - 4 | 269 | 94 | |
AK291381.1 | - | 3 - 4 | 269 | 94 | |
AK298714.1 | - | 3 - 4 | 251 | 94 | |
BC012620.1 | - | 3 - 4 | 258 | 94 | |
BC040471.1 | - | 3 - 4 | 250 | 94 | |
BE785872.1 | - | - | 231 | 94 | |
BG654545.1 | - | 3 - 4 | 242 | 94 |