Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007024.1 | NP_001007025.1 | 5 - 6 | 483 | 122 |
NM_001007025.1 | NP_001007026.1 | 5 - 6 | 461 | 122 | |
NM_004871.2 | NP_004862.1 | 5 - 6 | 467 | 122 | |
XM_005258070.4 | XP_005258127.1 | 5 - 6 | 708 | 122 | |
XM_005258071.4 | XP_005258128.1 | 5 - 6 | 701 | 122 | |
XM_005258072.2 | XP_005258129.1 | 5 - 6 | 495 | 122 | |
XM_017025374.1 | XP_016880863.1 | 5 - 6 | 711 | 122 | |
XM_017025375.1 | XP_016880864.1 | 5 - 6 | 470 | 122 | |
XM_017025376.1 | XP_016880865.1 | 5 - 6 | 452 | 122 | |
GenBank mRNA | AF073926.1 | - | 5 - 6 | 453 | 122 |
AK000606.1 | - | 5 - 6 | 467 | 122 | |
AK291381.1 | - | 5 - 6 | 467 | 122 | |
AK298714.1 | - | 5 - 6 | 449 | 122 | |
BC040471.1 | - | 5 - 6 | 448 | 122 | |
BG654545.1 | - | 5 - 6 | 440 | 122 |