Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021254.2 | NP_067077.1 | 4 - 5 | 896 | 75 |
NR_036552.1 | 3 - 4 | 823 | 75 | ||
GenBank mRNA | AF282851.1 | - | 4 - 5 | 644 | 75 |
AK000474.1 | - | 3 - 4 | 823 | 75 | |
AK094456.1 | - | 1 - 2 | 386 | 75 | |
AK097566.1 | - | - | 1797 | 75 | |
AK098577.1 | - | 4 - 5 | 896 | 75 | |
AK225697.1 | - | 4 - 5 | 874 | 75 | |
AK300970.1 | - | 3 - 4 | 706 | 75 | |
BC000709.2 | - | 4 - 5 | 649 | 75 | |
FJ649644.1 | - | 4 - 5 | 1824 | 75 |