Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206612.1 | NP_001193541.1 | 3 - 4 | 599 | 74 |
XM_017000993.1 | XP_016856482.1 | 3 - 4 | 558 | 74 | |
GenBank mRNA | BC108721.1 | - | 2 - 3 | 234 | 74 |
BC120961.1 | - | 3 - 4 | 326 | 74 | |
BC120962.1 | - | 3 - 4 | 326 | 74 |