Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145169.2 | NP_660152.1 | 2 - 3 | 276 | 94 |
NR_130112.1 | 3 - 4 | 299 | 94 | ||
NR_130113.1 | 3 - 4 | 299 | 94 | ||
NR_130114.1 | 3 - 4 | 290 | 94 | ||
GenBank mRNA | AF041429.1 | - | 2 - 3 | 210 | 94 |
AW964442.1 | - | 3 - 4 | 221 | 94 | |
BC018969.1 | - | 2 - 3 | 181 | 94 | |
BC028388.1 | - | 3 - 4 | 243 | 94 | |
BF223532.1 | - | 3 - 4 | 194 | 94 | |
BG706927.1 | - | 2 - 3 | 265 | 94 | |
DB476616.1 | - | 3 - 4 | 299 | 94 |