Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005192.3 | NP_005183.2 | 3 - 4 | 261 | 145 |
XM_011536353.2 | XP_011534655.1 | 3 - 4 | 277 | 145 | |
GenBank mRNA | AF213035.1 | - | 2 - 3 | 147 | 145 |
AF213036.1 | - | 2 - 3 | 147 | 145 | |
AF213039.1 | - | 2 - 3 | 147 | 145 | |
AF213040.1 | - | 2 - 3 | 147 | 145 | |
AF213041.1 | - | 2 - 3 | 147 | 145 | |
AF213046.1 | - | 2 - 3 | 147 | 145 | |
AF213050.1 | - | 2 - 3 | 147 | 145 | |
AF213052.1 | - | 2 - 3 | 147 | 145 | |
AK098653.1 | - | 3 - 4 | 261 | 145 | |
AY257474.1 | - | 2 - 3 | 147 | 145 | |
BT019451.1 | - | 2 - 3 | 147 | 145 | |
CR407666.1 | - | 2 - 3 | 147 | 145 | |
EF560750.1 | - | 3 - 4 | 187 | 145 | |
KP966095.1 | - | 2 - 3 | 178 | 145 | |
L25876.1 | - | 3 - 4 | 208 | 145 | |
L27711.1 | - | 3 - 4 | 198 | 145 | |
U02681.1 | - | 3 - 4 | 178 | 145 |