Assay Design



Gene Transcripts

Gene Symbol
WDR93
Entrez Gene ID
56964
Gene Name
WD repeat domain 93
Gene Aliases
-
Chromosome Location
Chr.15: 89690074 - 89744390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284395.1 NP_001271324.1 13 - 14 1557 63
NM_020212.1 NP_064597.1 13 - 14 1641 63
GenBank mRNA AK097525.1 - 13 - 14 1525 63
AK302032.1 - 7 - 8 1046 63
AL390084.1 - 5 - 6 625 63
AL390085.1 - 5 - 6 625 63
BC064626.1 - 13 - 14 1641 63

View Full Product Details