Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042471.1 | NP_001035936.1 | 6 - 7 | 817 | 85 |
XM_017024852.1 | XP_016880341.1 | 5 - 6 | 2747 | 85 | |
GenBank mRNA | AK000409.1 | - | 6 - 7 | 757 | 85 |
EU526753.1 | - | 5 - 6 | 686 | 85 |