Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042471.1 | NP_001035936.1 | 7 - 8 | 986 | 113 |
NM_001165958.1 | NP_001159430.1 | 8 - 9 | 1025 | 113 | |
NM_001165959.1 | NP_001159431.1 | 7 - 8 | 989 | 113 | |
NM_018530.2 | NP_061000.2 | 5 - 6 | 1052 | 113 | |
XM_011525004.1 | XP_011523306.1 | 8 - 9 | 2433 | 113 | |
XM_011525006.1 | XP_011523308.1 | 8 - 9 | 2424 | 113 | |
XM_011525007.2 | XP_011523309.1 | 8 - 9 | 1755 | 113 | |
XM_011525009.1 | XP_011523311.1 | 8 - 9 | 2113 | 113 | |
XM_011525011.1 | XP_011523313.1 | 8 - 9 | 1764 | 113 | |
XM_011525012.1 | XP_011523314.1 | 8 - 9 | 2104 | 113 | |
XM_011525013.1 | XP_011523315.1 | 9 - 10 | 1822 | 113 | |
XM_011525015.1 | XP_011523317.1 | 9 - 10 | 1813 | 113 | |
XM_017024848.1 | XP_016880337.1 | 8 - 9 | 2756 | 113 | |
XM_017024849.1 | XP_016880338.1 | 9 - 10 | 2465 | 113 | |
XM_017024850.1 | XP_016880339.1 | 7 - 8 | 2955 | 113 | |
XM_017024851.1 | XP_016880340.1 | 6 - 7 | 2940 | 113 | |
XM_017024852.1 | XP_016880341.1 | 6 - 7 | 2916 | 113 | |
GenBank mRNA | AF119884.1 | - | 5 - 6 | 1052 | 113 |
AF258572.1 | - | 1 - 2 | 1203 | 113 | |
AK000409.1 | - | 7 - 8 | 926 | 113 | |
AK057588.1 | - | 1 - 2 | 1635 | 113 | |
AK296607.1 | - | 7 - 8 | 974 | 113 | |
BC025682.1 | - | 6 - 7 | 902 | 113 | |
BX538068.1 | - | 7 - 8 | 689 | 113 | |
BX647700.1 | - | 6 - 7 | 4962 | 113 | |
CA488357.1 | - | 3 - 4 | 406 | 113 | |
EU526753.1 | - | 6 - 7 | 855 | 113 | |
EU526754.1 | - | 4 - 5 | 743 | 113 | |
EU526755.1 | - | 5 - 6 | 770 | 113 | |
EU526756.1 | - | 4 - 5 | 601 | 113 |