Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142294.1 | NP_001135766.1 | 4 - 5 | 1256 | 107 |
NM_001142295.1 | NP_001135767.1 | 4 - 5 | 1380 | 107 | |
NM_001142296.1 | NP_001135768.1 | 5 - 6 | 1432 | 107 | |
NM_015087.4 | NP_055902.1 | 4 - 5 | 1449 | 107 | |
XM_005266313.4 | XP_005266370.1 | 4 - 5 | 1374 | 107 | |
XM_005266314.3 | XP_005266371.1 | 5 - 6 | 1423 | 107 | |
XM_005266315.3 | XP_005266372.1 | 5 - 6 | 1277 | 107 | |
XM_005266317.3 | XP_005266374.1 | 4 - 5 | 1207 | 107 | |
XM_011535012.2 | XP_011533314.1 | 4 - 5 | 1360 | 107 | |
GenBank mRNA | AB011182.1 | - | 4 - 5 | 1223 | 107 |
AK001949.1 | - | 4 - 5 | 1213 | 107 | |
AK002207.1 | - | 3 - 4 | 400 | 107 | |
AK057764.1 | - | 5 - 6 | 769 | 107 | |
AY038359.1 | - | 3 - 4 | 1162 | 107 | |
AY123329.1 | - | 4 - 5 | 1223 | 107 | |
BC047083.1 | - | 4 - 5 | 1256 | 107 | |
BX648004.1 | - | 4 - 5 | 1345 | 107 | |
KU178563.1 | - | 3 - 4 | 1162 | 107 |