Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184993.1 | NP_001171922.1 | 31 - 32 | 4306 | 91 |
NM_031277.2 | NP_112567.2 | 31 - 32 | 4318 | 91 | |
XM_006719846.3 | XP_006719909.1 | 32 - 33 | 4759 | 91 | |
XM_006719849.2 | XP_006719912.1 | 32 - 33 | 4476 | 91 | |
XM_006719852.2 | XP_006719915.1 | 31 - 32 | 4354 | 91 | |
XM_011535152.2 | XP_011533454.1 | 36 - 37 | 6384 | 91 | |
XM_011535155.2 | XP_011533457.1 | 35 - 36 | 5120 | 91 | |
XM_011535156.2 | XP_011533458.1 | 36 - 37 | 6384 | 91 | |
XM_011535157.2 | XP_011533459.1 | 36 - 37 | 6372 | 91 | |
XM_011535158.2 | XP_011533460.1 | 35 - 36 | 6258 | 91 | |
XM_011535159.2 | XP_011533461.1 | 35 - 36 | 6237 | 91 | |
XM_011535160.2 | XP_011533462.1 | 35 - 36 | 6225 | 91 | |
XM_011535162.1 | XP_011533464.1 | 28 - 29 | 4000 | 91 | |
XM_011535163.1 | XP_011533465.1 | 23 - 24 | 3270 | 91 | |
XM_011535164.2 | XP_011533466.1 | 17 - 18 | 2166 | 91 | |
XM_017020676.1 | XP_016876165.1 | 32 - 33 | 4428 | 91 | |
GenBank mRNA | AK001907.1 | - | 11 - 12 | 1413 | 91 |
AK302435.1 | - | 31 - 32 | 4282 | 91 | |
BC064847.1 | - | 15 - 16 | 1969 | 91 | |
BC146822.1 | - | 32 - 33 | 4296 | 91 |