Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184993.1 | NP_001171922.1 | 33 - 34 | 4613 | 130 |
NM_031277.2 | NP_112567.2 | 33 - 34 | 4625 | 130 | |
XM_006719846.3 | XP_006719909.1 | 34 - 35 | 5066 | 130 | |
XM_006719849.2 | XP_006719912.1 | 34 - 35 | 4783 | 130 | |
XM_006719852.2 | XP_006719915.1 | 33 - 34 | 4661 | 130 | |
XM_011535152.2 | XP_011533454.1 | 38 - 39 | 6691 | 130 | |
XM_011535155.2 | XP_011533457.1 | 37 - 38 | 5427 | 130 | |
XM_011535156.2 | XP_011533458.1 | 38 - 39 | 6691 | 130 | |
XM_011535157.2 | XP_011533459.1 | 38 - 39 | 6679 | 130 | |
XM_011535158.2 | XP_011533460.1 | 37 - 38 | 6565 | 130 | |
XM_011535159.2 | XP_011533461.1 | 37 - 38 | 6544 | 130 | |
XM_011535160.2 | XP_011533462.1 | 37 - 38 | 6532 | 130 | |
XM_011535162.1 | XP_011533464.1 | 30 - 31 | 4307 | 130 | |
XM_011535163.1 | XP_011533465.1 | 25 - 26 | 3577 | 130 | |
XM_011535164.2 | XP_011533466.1 | 19 - 20 | 2473 | 130 | |
XM_017020676.1 | XP_016876165.1 | 34 - 35 | 4735 | 130 | |
GenBank mRNA | AK001907.1 | - | 13 - 14 | 1720 | 130 |
AK302435.1 | - | 33 - 34 | 4589 | 130 | |
BC064847.1 | - | 17 - 18 | 2276 | 130 | |
BC146822.1 | - | 34 - 35 | 4603 | 130 |