Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322903.1 | NP_001309832.1 | 4 - 5 | 484 | 86 |
NM_001322904.1 | NP_001309833.1 | 4 - 5 | 454 | 86 | |
NM_001322905.1 | NP_001309834.1 | 4 - 5 | 417 | 86 | |
NM_001322906.1 | NP_001309835.1 | 4 - 5 | 411 | 86 | |
NM_001322907.1 | NP_001309836.1 | 5 - 6 | 612 | 86 | |
NM_001322908.1 | NP_001309837.1 | 5 - 6 | 535 | 86 | |
NM_001322909.1 | NP_001309838.1 | 4 - 5 | 402 | 86 | |
NM_001322910.1 | NP_001309839.1 | 4 - 5 | 509 | 86 | |
NM_130811.3 | NP_570824.1 | 3 - 4 | 357 | 86 | |
XM_005260808.4 | XP_005260865.1 | 3 - 4 | 435 | 86 | |
XM_017028023.1 | XP_016883512.1 | 3 - 4 | 461 | 86 | |
GenBank mRNA | AK098347.1 | - | 4 - 5 | 230 | 86 |
AK223617.1 | - | 3 - 4 | 319 | 86 | |
AK314359.1 | - | 3 - 4 | 268 | 86 | |
BI552777.1 | - | 3 - 4 | 330 | 86 | |
BI604120.1 | - | 3 - 4 | 334 | 86 | |
BX647443.1 | - | 3 - 4 | 229 | 86 | |
D21267.1 | - | 3 - 4 | 319 | 86 | |
DA072367.1 | - | 3 - 4 | 357 | 86 | |
DA162693.1 | - | 3 - 4 | 461 | 86 | |
L19761.1 | - | 3 - 4 | 202 | 86 |