Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005522.4 | NP_005513.1 | 1 - 2 | 750 | 77 |
NM_153620.2 | NP_705873.2 | 2 - 3 | 547 | 77 | |
GenBank mRNA | AK308067.1 | - | 2 - 3 | 495 | 77 |
AK313514.1 | - | 1 - 2 | 741 | 77 | |
BC032547.2 | - | 1 - 2 | 718 | 77 | |
S79871.1 | - | 2 - 3 | 447 | 77 | |
S79910.1 | - | 1 - 2 | 650 | 77 | |
U10421.1 | - | 1 - 2 | 656 | 77 |