Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000970.4 | NP_000961.2 | 4 - 5 | 537 | 70 |
NM_001024662.2 | NP_001019833.1 | 4 - 5 | 713 | 70 | |
NM_001320137.1 | NP_001307066.1 | 4 - 5 | 718 | 70 | |
NM_001320138.1 | NP_001307067.1 | 4 - 5 | 752 | 70 | |
NM_001320139.1 | NP_001307068.1 | 4 - 5 | 599 | 70 | |
NM_001320140.1 | NP_001307069.1 | 4 - 5 | 761 | 70 | |
NM_001320141.1 | NP_001307070.1 | 6 - 7 | 826 | 70 | |
NM_001320142.1 | NP_001307071.1 | 4 - 5 | 473 | 70 | |
XM_017019781.1 | XP_016875270.1 | 6 - 7 | 1861 | 70 | |
XM_017019782.1 | XP_016875271.1 | 6 - 7 | 1022 | 70 | |
GenBank mRNA | AK299470.1 | - | 5 - 6 | 467 | 70 |
AK311863.1 | - | 4 - 5 | 501 | 70 | |
AY320410.1 | - | 1 - 2 | 57 | 70 | |
BC004138.2 | - | 1 - 1 | 513 | 70 | |
BC013863.1 | - | 1 - 1 | 487 | 70 | |
BC020679.1 | - | 1 - 1 | 501 | 70 | |
BC022444.1 | - | 4 - 5 | 668 | 70 | |
BC031009.1 | - | 1 - 1 | 501 | 70 | |
BC032299.1 | - | 1 - 1 | 498 | 70 | |
BC071912.1 | - | 1 - 1 | 490 | 70 | |
BC104824.1 | - | 4 - 5 | 513 | 70 | |
BC104826.1 | - | 4 - 5 | 513 | 70 | |
BG030827.1 | - | 1 - 1 | 263 | 70 | |
CD049618.1 | - | 4 - 5 | 683 | 70 | |
D17554.1 | - | 4 - 5 | 503 | 70 | |
S71022.1 | - | 3 - 4 | 347 | 70 | |
X69391.1 | - | 4 - 5 | 508 | 70 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AK126570.1 | - | - | 1249 | 70 |