Assay Design



Gene Transcripts

Gene Symbol
RPL6
Entrez Gene ID
6128
Gene Name
ribosomal protein L6
Gene Aliases
L6, SHUJUN-2, TAXREB107, TXREB1
Chromosome Location
Chr.12: 112405181 - 112418850 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320141.1 NP_001307070.1 3 - 4 348 82
XM_017019781.1 XP_016875270.1 3 - 4 1383 82
XM_017019782.1 XP_016875271.1 3 - 4 544 82
GenBank mRNA BG722198.1 - 3 - 4 348 82
HY038019.1 - 3 - 4 348 82

View Full Product Details