Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128431.2 | NP_001121903.1 | 6 - 7 | 1110 | 76 |
NM_001135153.1 | NP_001128625.1 | 6 - 7 | 1151 | 76 | |
NM_001135154.1 | NP_001128626.1 | 6 - 7 | 1110 | 76 | |
NM_015359.4 | NP_056174.2 | 6 - 7 | 1110 | 76 | |
XM_005273465.2 | XP_005273522.1 | 7 - 8 | 1198 | 76 | |
XM_005273466.4 | XP_005273523.1 | 6 - 7 | 1182 | 76 | |
XM_006716323.2 | XP_006716386.1 | 8 - 9 | 1396 | 76 | |
XM_006716324.2 | XP_006716387.1 | 7 - 8 | 1187 | 76 | |
XM_011544478.2 | XP_011542780.1 | 7 - 8 | 1262 | 76 | |
XM_017013292.1 | XP_016868781.1 | 7 - 8 | 1278 | 76 | |
XM_017013293.1 | XP_016868782.1 | 8 - 9 | 1396 | 76 | |
XM_017013294.1 | XP_016868783.1 | 8 - 9 | 1305 | 76 | |
XM_017013295.1 | XP_016868784.1 | 6 - 7 | 1315 | 76 | |
GenBank mRNA | AK172810.1 | - | 6 - 7 | 1081 | 76 |
BC015770.1 | - | 6 - 7 | 1072 | 76 | |
D31887.1 | - | 6 - 7 | 1054 | 76 | |
KU178568.1 | - | 5 - 6 | 935 | 76 | |
KU178569.1 | - | 5 - 6 | 935 | 76 | |
KU178570.1 | - | 5 - 6 | 935 | 76 | |
KU178571.1 | - | 5 - 6 | 935 | 76 |