Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039937.1 | NP_001035026.1 | 14 - 15 | 1850 | 67 |
NM_001306091.1 | NP_001293020.1 | 15 - 16 | 2626 | 67 | |
NM_012141.2 | NP_036273.1 | 13 - 14 | 2342 | 67 | |
XM_011535040.2 | XP_011533342.1 | 13 - 14 | 2439 | 67 | |
XM_011535041.1 | XP_011533343.1 | 12 - 13 | 2255 | 67 | |
XM_017020540.1 | XP_016876029.1 | 12 - 13 | 1800 | 67 | |
GenBank mRNA | AF097645.1 | - | 13 - 14 | 2204 | 67 |
AF141326.1 | - | 7 - 8 | 913 | 67 | |
AK074946.1 | - | 11 - 12 | 1432 | 67 | |
AK096696.1 | - | 10 - 11 | 1562 | 67 | |
AK128795.1 | - | 11 - 12 | 1497 | 67 | |
AK304218.1 | - | 12 - 13 | 2144 | 67 | |
AL117626.1 | - | 4 - 5 | 497 | 67 | |
AL833524.1 | - | 14 - 15 | 1918 | 67 | |
BC013358.2 | - | 13 - 14 | 2220 | 67 | |
BC039829.1 | - | 13 - 14 | 2216 | 67 |