Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039937.1 | NP_001035026.1 | 4 - 5 | 460 | 135 |
NM_001306091.1 | NP_001293020.1 | 3 - 4 | 952 | 135 | |
NM_012141.2 | NP_036273.1 | 3 - 4 | 952 | 135 | |
XM_011535040.2 | XP_011533342.1 | 3 - 4 | 1049 | 135 | |
XM_011535041.1 | XP_011533343.1 | 3 - 4 | 1049 | 135 | |
XM_017020540.1 | XP_016876029.1 | 3 - 4 | 505 | 135 | |
GenBank mRNA | AF097645.1 | - | 3 - 4 | 814 | 135 |
AK128795.1 | - | 1 - 2 | 107 | 135 | |
AK296801.1 | - | 3 - 4 | 849 | 135 | |
AK303869.1 | - | 3 - 4 | 443 | 135 | |
AK304218.1 | - | 3 - 4 | 849 | 135 | |
AL833524.1 | - | 4 - 5 | 528 | 135 | |
BC013358.2 | - | 3 - 4 | 830 | 135 | |
BC039829.1 | - | 3 - 4 | 826 | 135 |