Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145666.1 | NP_001139138.1 | 10 - 11 | 1847 | 72 |
NM_001145667.1 | NP_001139139.1 | 11 - 12 | 1880 | 72 | |
NM_012201.5 | NP_036333.2 | 11 - 12 | 1880 | 72 | |
NR_027264.1 | 11 - 12 | 1880 | 72 | ||
NR_027265.1 | 6 - 7 | 877 | 72 | ||
GenBank mRNA | AK025457.1 | - | 8 - 9 | 1273 | 72 |
AK026972.1 | - | 2 - 3 | 239 | 72 | |
AK027032.1 | - | 11 - 12 | 1841 | 72 | |
AK304156.1 | - | 10 - 11 | 1817 | 72 | |
BC031836.1 | - | 6 - 7 | 831 | 72 | |
BC060822.1 | - | 11 - 12 | 1851 | 72 | |
U64791.1 | - | 11 - 12 | 1833 | 72 |