Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145666.1 | NP_001139138.1 | 5 - 6 | 1070 | 97 |
NM_001145667.1 | NP_001139139.1 | 6 - 7 | 1103 | 97 | |
NM_012201.5 | NP_036333.2 | 6 - 7 | 1103 | 97 | |
NR_027264.1 | 6 - 7 | 1103 | 97 | ||
GenBank mRNA | AK027032.1 | - | 6 - 7 | 1064 | 97 |
AK304156.1 | - | 5 - 6 | 1040 | 97 | |
BC060822.1 | - | 6 - 7 | 1074 | 97 | |
U28811.1 | - | 6 - 7 | 1076 | 97 | |
U64791.1 | - | 6 - 7 | 1056 | 97 |