Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098272.2 | NP_001091742.1 | 7 - 8 | 1300 | 80 |
NM_001324219.1 | NP_001311148.1 | 6 - 7 | 1287 | 80 | |
NM_001324220.1 | NP_001311149.1 | 7 - 8 | 1320 | 80 | |
NM_001324222.1 | NP_001311151.1 | 6 - 7 | 1174 | 80 | |
NM_001324223.1 | NP_001311152.1 | 5 - 6 | 1115 | 80 | |
NM_001324224.1 | NP_001311153.1 | 6 - 7 | 1194 | 80 | |
NM_002130.7 | NP_002121.4 | 6 - 7 | 1241 | 80 | |
XM_011514036.2 | XP_011512338.1 | 4 - 5 | 1106 | 80 | |
XM_017009412.1 | XP_016864901.1 | 7 - 8 | 1380 | 80 | |
GenBank mRNA | AK095492.1 | - | 7 - 8 | 1108 | 80 |
AK315593.1 | - | 7 - 8 | 1228 | 80 | |
BC000297.2 | - | 7 - 8 | 1179 | 80 | |
BC083514.1 | - | 6 - 7 | 1139 | 80 | |
BT007302.1 | - | 5 - 6 | 1074 | 80 | |
L25798.1 | - | 5 - 6 | 1095 | 80 |