Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032865.5 | NP_116254.4 | 3 - 4 | 1027 | 75 |
XM_017025236.1 | XP_016880725.1 | 3 - 4 | 1106 | 75 | |
XM_017025237.1 | XP_016880726.1 | 3 - 4 | 1106 | 75 | |
GenBank mRNA | AF370421.1 | - | 3 - 4 | 1024 | 74 |
AF417488.1 | - | 3 - 4 | 989 | 75 | |
AK027856.1 | - | 1 - 2 | 377 | 75 | |
BC018706.1 | - | 2 - 3 | 473 | 75 |