Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181787.2 | NP_861452.2 | 9 - 10 | 1127 | 152 |
XM_005250894.4 | XP_005250951.2 | 9 - 10 | 1340 | 152 | |
XM_005250895.3 | XP_005250952.1 | 8 - 9 | 1008 | 152 | |
XM_011516984.2 | XP_011515286.1 | 8 - 9 | 1007 | 152 | |
XM_017013352.1 | XP_016868841.1 | 7 - 8 | 1078 | 152 | |
XM_017013354.1 | XP_016868843.1 | 8 - 9 | 1033 | 152 | |
XM_017013355.1 | XP_016868844.1 | 8 - 9 | 1026 | 152 | |
XM_017013356.1 | XP_016868845.1 | 7 - 8 | 879 | 152 | |
GenBank mRNA | AK123618.1 | - | 1 - 2 | 122 | 152 |
AK123682.1 | - | 9 - 10 | 1081 | 152 | |
AL832859.1 | - | 4 - 5 | 442 | 152 | |
BC110870.1 | - | 9 - 10 | 1055 | 152 | |
BC126193.1 | - | 9 - 10 | 1023 | 152 | |
BC130576.1 | - | 9 - 10 | 1023 | 152 | |
BX538048.1 | - | 9 - 10 | 1075 | 152 | |
BX538174.1 | - | 9 - 10 | 1079 | 152 |