Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006948.4 | NP_008879.3 | 2 - 3 | 431 | 79 |
GenBank mRNA | AK223282.1 | - | 2 - 3 | 389 | 79 |
AK312396.1 | - | 2 - 3 | 386 | 79 | |
BC036370.2 | - | 2 - 3 | 393 | 79 | |
U04735.1 | - | 2 - 3 | 398 | 79 |