Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006576.3 | NP_006567.3 | 4 - 5 | 477 | 71 |
XM_017018710.1 | XP_016874199.1 | 5 - 6 | 543 | 71 | |
XM_017018711.1 | XP_016874200.1 | 5 - 6 | 3289 | 71 | |
XM_017018712.1 | XP_016874201.1 | 5 - 6 | 3232 | 71 | |
XM_017018713.1 | XP_016874202.1 | 5 - 6 | 2892 | 71 | |
GenBank mRNA | AF041449.1 | - | 4 - 5 | 477 | 71 |
AK314362.1 | - | 4 - 5 | 879 | 71 | |
BC111730.1 | - | 5 - 6 | 581 | 71 |