Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023012.5 | NP_075388.2 | 9 - 10 | 1285 | 152 |
NR_036434.1 | 10 - 11 | 1366 | 152 | ||
NR_036435.1 | 10 - 11 | 1385 | 152 | ||
NR_036436.1 | 9 - 10 | 1307 | 152 | ||
XM_005253601.1 | XP_005253658.1 | 9 - 10 | 1288 | 152 | |
XM_005253602.2 | XP_005253659.1 | 9 - 10 | 1259 | 152 | |
XM_005253604.1 | XP_005253661.1 | 10 - 11 | 1370 | 152 | |
XM_011538689.1 | XP_011536991.1 | 12 - 13 | 1543 | 152 | |
XM_017019831.1 | XP_016875320.1 | 10 - 11 | 1365 | 152 | |
XM_017019832.1 | XP_016875321.1 | 12 - 13 | 1521 | 152 | |
XM_017019833.1 | XP_016875322.1 | 10 - 11 | 1312 | 152 | |
GenBank mRNA | AB212664.1 | - | 9 - 10 | 1271 | 152 |
AK001883.1 | - | 9 - 10 | 1245 | 152 | |
AK023985.1 | - | 6 - 7 | 875 | 152 | |
AK056114.1 | - | 10 - 11 | 1307 | 152 | |
BC008684.1 | - | 9 - 10 | 1195 | 152 | |
BC067773.1 | - | 9 - 10 | 1255 | 152 | |
BX640711.1 | - | 9 - 10 | 1231 | 152 | |
BX647884.1 | - | 9 - 10 | 1274 | 152 |