Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261412.1 | NP_001248341.1 | 8 - 9 | 938 | 60 |
NM_001261413.1 | NP_001248342.1 | 7 - 8 | 932 | 60 | |
NM_006400.4 | NP_006391.1 | 7 - 8 | 947 | 60 | |
XM_011537761.2 | XP_011536063.1 | 8 - 9 | 899 | 60 | |
GenBank mRNA | AK055682.1 | - | 7 - 8 | 810 | 60 |
AK096242.1 | - | 9 - 10 | 806 | 60 | |
AK098794.1 | - | 8 - 9 | 792 | 60 | |
AK222693.1 | - | 7 - 8 | 810 | 60 | |
AK292364.1 | - | 9 - 10 | 825 | 60 | |
AK314705.1 | - | 7 - 8 | 789 | 60 | |
AY189155.1 | - | 8 - 9 | 670 | 60 | |
BC000718.2 | - | 7 - 8 | 796 | 60 | |
BC009468.1 | - | 7 - 8 | 722 | 60 | |
BC014083.2 | - | 7 - 8 | 770 | 60 | |
FJ224315.1 | - | 7 - 8 | 757 | 60 | |
U50733.1 | - | 7 - 8 | 757 | 60 |