Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002827.3 | NP_002818.1 | 1 - 2 | 265 | 97 |
GenBank mRNA | AK290638.1 | - | 1 - 2 | 232 | 97 |
AU117677.1 | - | 1 - 2 | 232 | 97 | |
BC015660.2 | - | 1 - 2 | 194 | 97 | |
BC018164.1 | - | 1 - 2 | 175 | 97 | |
BT006752.1 | - | 1 - 2 | 58 | 97 | |
M31724.1 | - | 1 - 2 | 148 | 97 | |
M33689.1 | - | 1 - 2 | 130 | 97 |