Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278618.1 | NP_001265547.1 | 6 - 7 | 983 | 71 |
NM_002827.3 | NP_002818.1 | 7 - 8 | 1074 | 71 | |
GenBank mRNA | AK290638.1 | - | 7 - 8 | 1041 | 71 |
AK299830.1 | - | 6 - 7 | 921 | 71 | |
AK316563.1 | - | 7 - 8 | 1016 | 71 | |
BC015660.2 | - | 7 - 8 | 1003 | 71 | |
BC018164.1 | - | 7 - 8 | 984 | 71 | |
BT006752.1 | - | 7 - 8 | 867 | 71 | |
M31724.1 | - | 7 - 8 | 957 | 71 | |
M33689.1 | - | 7 - 8 | 939 | 71 |