Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278618.1 | NP_001265547.1 | 7 - 8 | 1204 | 95 |
NM_002827.3 | NP_002818.1 | 8 - 9 | 1295 | 95 | |
GenBank mRNA | AK290638.1 | - | 8 - 9 | 1262 | 95 |
AK299830.1 | - | 7 - 8 | 1142 | 95 | |
AK316563.1 | - | 8 - 9 | 1237 | 95 | |
BC015660.2 | - | 8 - 9 | 1224 | 95 | |
BC018164.1 | - | 8 - 9 | 1205 | 95 | |
BT006752.1 | - | 8 - 9 | 1088 | 95 | |
M31724.1 | - | 8 - 9 | 1178 | 95 | |
M33689.1 | - | 8 - 9 | 1160 | 95 |