Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020312.3 | NP_064708.1 | 4 - 5 | 596 | 61 |
GenBank mRNA | AF161444.1 | - | 4 - 5 | 530 | 61 |
AK290627.1 | - | 4 - 5 | 526 | 61 | |
AK295793.1 | - | 4 - 5 | 539 | 61 | |
BC001478.2 | - | 2 - 3 | 171 | 61 | |
BC029341.1 | - | 4 - 5 | 530 | 61 | |
BC054340.1 | - | 4 - 5 | 530 | 61 | |
BC064946.1 | - | 4 - 5 | 531 | 61 |