Assay Design



Gene Transcripts

Gene Symbol
WDR27
Entrez Gene ID
253769
Gene Name
WD repeat domain 27
Gene Aliases
-
Chromosome Location
Chr.6: 169426420 - 169702071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202550.1 NP_001189479.1 14 - 15 1882 93
NM_182552.4 NP_872358.4 17 - 18 2263 93
XM_011535682.2 XP_011533984.1 18 - 19 2453 93
XM_011535684.2 XP_011533986.1 18 - 19 2453 93
XM_011535685.2 XP_011533987.1 18 - 19 2453 93
XM_011535687.2 XP_011533989.1 18 - 19 2453 93
XM_011535688.2 XP_011533990.1 18 - 19 2453 93
XM_011535689.2 XP_011533991.1 18 - 19 2453 93
XM_011535690.2 XP_011533992.1 18 - 19 2453 93
XM_011535691.2 XP_011533993.1 18 - 19 2453 93
XM_011535692.2 XP_011533994.1 18 - 19 2453 93
XM_011535693.2 XP_011533995.1 18 - 19 2453 93
XM_011535694.2 XP_011533996.1 18 - 19 2453 93
XM_017010658.1 XP_016866147.1 18 - 19 2453 93
XM_017010659.1 XP_016866148.1 18 - 19 2453 93
XM_017010660.1 XP_016866149.1 18 - 19 2453 93
XM_017010661.1 XP_016866150.1 18 - 19 2453 93
XM_017010662.1 XP_016866151.1 18 - 19 2453 93
XM_017010663.1 XP_016866152.1 18 - 19 2453 93
XM_017010664.1 XP_016866153.1 18 - 19 2453 93
XM_017010665.1 XP_016866154.1 18 - 19 2453 93
XM_017010666.1 XP_016866155.1 18 - 19 2453 93
XM_017010667.1 XP_016866156.1 18 - 19 2453 93
XM_017010668.1 XP_016866157.1 18 - 19 2453 93
XM_017010669.1 XP_016866158.1 12 - 13 1698 93
XM_017010670.1 XP_016866159.1 18 - 19 2453 93
XM_017010671.1 XP_016866160.1 18 - 19 2453 93
GenBank mRNA AK131435.1 - 8 - 9 2644 93
BC131633.1 - 15 - 16 1405 93
BX648854.1 - 13 - 14 1258 93

View Full Product Details