Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202550.1 | NP_001189479.1 | 18 - 19 | 2356 | 64 |
NM_182552.4 | NP_872358.4 | 21 - 22 | 2737 | 64 | |
XM_011535682.2 | XP_011533984.1 | 22 - 23 | 2927 | 64 | |
XM_011535684.2 | XP_011533986.1 | 22 - 23 | 2927 | 64 | |
XM_011535685.2 | XP_011533987.1 | 22 - 23 | 2927 | 64 | |
XM_011535687.2 | XP_011533989.1 | 22 - 23 | 2927 | 64 | |
XM_011535688.2 | XP_011533990.1 | 22 - 23 | 2927 | 64 | |
XM_011535689.2 | XP_011533991.1 | 22 - 23 | 2927 | 64 | |
XM_011535690.2 | XP_011533992.1 | 22 - 23 | 2927 | 64 | |
XM_011535691.2 | XP_011533993.1 | 22 - 23 | 2927 | 64 | |
XM_011535692.2 | XP_011533994.1 | 22 - 23 | 2927 | 64 | |
XM_017010658.1 | XP_016866147.1 | 22 - 23 | 2927 | 64 | |
XM_017010659.1 | XP_016866148.1 | 22 - 23 | 2927 | 64 | |
XM_017010660.1 | XP_016866149.1 | 22 - 23 | 2927 | 64 | |
XM_017010661.1 | XP_016866150.1 | 22 - 23 | 2927 | 64 | |
XM_017010662.1 | XP_016866151.1 | 22 - 23 | 2927 | 64 | |
XM_017010669.1 | XP_016866158.1 | 16 - 17 | 2172 | 64 | |
XM_017010670.1 | XP_016866159.1 | 21 - 22 | 2793 | 64 | |
GenBank mRNA | BC131633.1 | - | 19 - 20 | 1879 | 64 |
BX648854.1 | - | 16 - 17 | 1598 | 64 |