Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080463.1 | NP_001073932.1 | 18 - 19 | 2847 | 147 |
NM_001377.2 | NP_001368.2 | 18 - 19 | 2847 | 147 | |
XM_006718903.2 | XP_006718966.1 | 18 - 19 | 2847 | 147 | |
XM_017018291.1 | XP_016873780.1 | 18 - 19 | 2847 | 147 | |
XM_017018292.1 | XP_016873781.1 | 15 - 16 | 2101 | 147 | |
XM_017018293.1 | XP_016873782.1 | 18 - 19 | 2847 | 147 | |
GenBank mRNA | AB290167.1 | - | 18 - 19 | 2901 | 147 |
AK125524.1 | - | 18 - 19 | 2847 | 147 |