Assay Design
Gene Transcripts
- Gene Symbol
- PATE1
- Entrez Gene ID
-
160065
- Gene Name
- prostate and testis expressed 1
- Gene Aliases
-
PATE
- Chromosome Location
-
Chr.11: 125746293 - 125749848 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC107044.2 |
- |
2 - 3 |
90 |
81 |
View Full Product Details