Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020936.2 | NP_065987.1 | 7 - 8 | 815 | 74 |
NR_135104.1 | 7 - 8 | 815 | 74 | ||
NR_135105.1 | 7 - 8 | 815 | 74 | ||
XM_011538608.2 | XP_011536910.1 | 7 - 8 | 803 | 74 | |
XM_017019710.1 | XP_016875199.1 | 7 - 8 | 800 | 74 | |
XM_017019711.1 | XP_016875200.1 | 6 - 7 | 855 | 74 | |
XM_017019712.1 | XP_016875201.1 | 7 - 8 | 653 | 74 | |
XM_017019713.1 | XP_016875202.1 | 7 - 8 | 830 | 74 | |
XM_017019714.1 | XP_016875203.1 | 8 - 9 | 988 | 74 | |
XM_017019715.1 | XP_016875204.1 | 7 - 8 | 801 | 74 | |
XM_017019716.1 | XP_016875205.1 | 7 - 8 | 800 | 74 | |
XM_017019717.1 | XP_016875206.1 | 7 - 8 | 698 | 74 | |
XM_017019718.1 | XP_016875207.1 | 5 - 6 | 1116 | 74 | |
XM_017019719.1 | XP_016875208.1 | 7 - 8 | 707 | 74 | |
XM_017019720.1 | XP_016875209.1 | 5 - 6 | 1785 | 74 | |
XM_017019721.1 | XP_016875210.1 | 7 - 8 | 801 | 74 | |
GenBank mRNA | AB046815.1 | - | 2 - 3 | 2642 | 74 |
AK131442.1 | - | 4 - 5 | 1054 | 74 | |
AK301043.1 | - | 7 - 8 | 778 | 74 | |
AK307384.1 | - | 7 - 8 | 791 | 74 | |
AL833756.1 | - | 5 - 6 | 1852 | 74 | |
BC030020.2 | - | 7 - 8 | 759 | 74 |