Assay Design
Gene Transcripts
- Gene Symbol
- CHST5
- Entrez Gene ID
-
23563
- Gene Name
- carbohydrate sulfotransferase 5
- Gene Aliases
-
I-GlcNAc-6-ST, I-GlcNAc6ST, glcNAc6ST-3, gn6st-3, hIGn6ST
- Chromosome Location
-
Chr.16: 75528530 - 75535170 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_024533.4 |
NP_078809.2 |
1 - 2 |
48 |
76 |
GenBank mRNA |
BX282540.1 |
- |
1 - 2 |
125 |
76 |
View Full Product Details