Assay Design



Gene Transcripts

Gene Symbol
CKMT2
Entrez Gene ID
1160
Gene Name
creatine kinase, mitochondrial 2
Gene Aliases
SMTCK
Chromosome Location
Chr.5: 81233320 - 81266398 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001825.2 NP_001816.2 2 - 3 222 72
GenBank mRNA DA826840.1 - 2 - 3 218 72
J05401.1 - 1 - 2 178 72

View Full Product Details