Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256404.1 | NP_001243333.1 | 12 - 13 | 2435 | 59 |
NM_198459.3 | NP_940861.3 | 9 - 10 | 1895 | 59 | |
GenBank mRNA | BX647950.1 | - | 9 - 10 | 1896 | 59 |
BX649075.1 | - | 9 - 10 | 1021 | 59 | |
CR749576.1 | - | 12 - 13 | 2359 | 59 |