Assay Design
Gene Transcripts
- Gene Symbol
- NOM1
- Entrez Gene ID
-
64434
- Gene Name
- nucleolar protein with MIF4G domain 1
- Gene Aliases
-
C7orf3, PPP1R113, SGD1
- Chromosome Location
-
Chr.7: 156949723 - 156973182 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138400.1 |
NP_612409.1 |
1 - 2 |
1003 |
84 |
GenBank mRNA |
AY576779.1 |
- |
1 - 2 |
1003 |
84 |
View Full Product Details